
2-Naphthalenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Codice catalogo TCIAN0018-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:75.0 to 79.0 deg-C
- Solubility in Toluene:very faint turbidity
Specification Test Results
About this item
:
:226.68 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:71…72 °C
EINECS:202-219-9
UN:3261
ADR:8,III
:GHS05
CAS Number:93-11-8
Formula:C₁₀H₇ClO₂S
MDL Number:MFCD00004087