Neutral red
Glass bottle, 25 g
Codice catalogo TCIAN0315-25G
Nome del fornitore: TCI
Jump to:
Product Details & Documents
Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
- CAS Number:553-24-2
- Formula:C₁₅H₁₇ClN₄
- MDL Number:MFCD00012651
- Molecular Weight:288.78 g/mol
- Merck Index:14,06488
- Melting Pt:290°C
- Storage Temperature:Ambient
- EINECS:209-035-8
Specifications
Specification Test Results
- Appearance:Dark green to Dark blue to Black powder to crystal
- Absorbance:min. 0.500 (10 mg/L, pH 6.4, 523.0 to 527.0 nm) (calcd.on dried substance)
- Drying loss:max. 20.0 %
- Transition interval (pH):(red)6.8 to 8.0 (yellowish orange)
Specifications
- Pack type:Glass bottle
- Pk:25 g



