Order Entry
Italy
ContactUsLinkComponent
3-(Methylthio)propyl isothiocyanate ≥97.0% (by GC)
Irritant
3-(Methylthio)propyl isothiocyanate ≥97.0% (by GC)
Catalog # TCIAM2027-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-(Methylthio)propyl isothiocyanate ≥97.0% (by GC)
Catalog # TCIAM2027-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :147.27 g/mol
Boiling Pt:122°C
EINECS:208-020-3
 :GHS07
CAS Number:505-79-3
Formula:C₅H₉NS₂
MDL Number:MFCD00209521