
1-Hexyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:304680-35-1
:
Formula:C₁₀H₁₉F₆N₂P
MDL Number:MFCD03093296
:312.24 g/mol
Density:1,28 g/cm³ (20 °C)
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (Ion exchange titration):min. 98.0 %
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together