4,4'-Difluorobenzhydrol ≥98.0% (by GC)
Catalog # TCIAD1795-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:46.0 to 49.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
About this item
:
:220.22 g/mol
Boiling Pt:154°C
Melting Pt:47 °C
Density:1 g/cm³ (20 °C)
EINECS:206-671-8
CAS Number:365-24-2
Formula:C₁₃H₁₀F₂O
MDL Number:MFCD00000357