
1-Ethyl-3-methylimidazolium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:65039-08-9
:
Formula:C₆H₁₁BrN₂
MDL Number:MFCD03427610
:191.07 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together