
m-Toluonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:620-22-4
:
Formula:C₈H₇N
MDL Number:MFCD00001808
:117.15 g/mol
Boiling Pt:210°C
Melting Pt:–0.5 °C
Density:0,976 g/cm³ (20 °C)
Flash Pt:86°C
EINECS:210-631-5
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together