

4-Chloro-N-methylaniline ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:932-96-7
:
Formula:C₇H₈ClN
MDL Number:MFCD00000614
:141.6 g/mol
Boiling Pt:142°C
Density:1,169 g/cm³ (12 °C)
Flash Pt:51°C
EINECS:213-262-8
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together