4,4'-Dichlorobenzophenone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90-98-2
:
Formula:C₁₃H₈Cl₂O
MDL Number:MFCD00000623
:251.11 g/mol
EINECS:202-030-1
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:146.0 to 149.0 deg-C
- Solubility in hot Acetone:within almost transparency
Specification Test Results
Frequently Bought Together