Order Entry
Italy
ContactUsLinkComponent
2-Bromo-5-chlorobenzotrifluoride ≥98.0% (by GC)
 
2-Bromo-5-chlorobenzotrifluoride ≥98.0% (by GC)
  TCIAB3366-5G
 :  TCI
 :  
undefined
 
2-Bromo-5-chlorobenzotrifluoride ≥98.0% (by GC)
  TCIAB3366-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :259.45 g/mol
 :198°C
 :19°C
 :99°C
 :GHS07
 :344-65-0
 :C₇H₃BrClF₃
 :MFCD00010308