


4,4'-Diaminodiphenyl ether ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:101-80-4
:
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD00007863
:200.24 g/mol
Boiling Pt:389 °C (1013 hPa)
Melting Pt:184 °C
Flash Pt:218 °C
EINECS:202-977-0
UN:3077
ADR:9,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:190.0 to 194.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together