Order Entry
Italy
ContactUsLinkComponent
3-(Methylthio)aniline ≥98.0% (by GC, titration analysis)
Irritant
3-(Methylthio)aniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAM1673-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-(Methylthio)aniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAM1673-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :139.22 g/mol
Boiling Pt:165°C
Density:1,1464 g/cm³ (20 °C)
EINECS:217-232-5
ADR:6.1,III
 :GHS07
CAS Number:1783-81-9
Formula:C₇H₉NS
MDL Number:MFCD00007793