
2-Chloro-4,6-difluoroaniline ≥98.0% (by GC, titration analysis)
: TCI
:36556-56-6
:
:C₆H₄ClF₂N
:MFCD00142892
:163.55 g/mol
:6.1,III
:GHS07
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together