
2-(Methylthio)aniline ≥97.0% (by GC)
Catalog # TCIAM1159-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:139.22 g/mol
Boiling Pt:242°C
Density:1,1333 g/cm³ (17 °C)
EINECS:221-055-9
UN:2810
ADR:6.1,III
:GHS06
CAS Number:2987-53-3
Formula:C₇H₉NS
MDL Number:MFCD00007708