
Ethyl hydrogen adipate ≥97.0% (by GC, titration analysis)
: TCI
:626-86-8
:
:C₈H₁₄O₄
:MFCD00004419
:174.2 g/mol
:286 °C (1013 hPa)
:27.4 °C
:1,0622 g/cm³ (20 °C)
:210-966-7
:GHS07
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Frequently Bought Together