

1,6-Bis(dimethylamino)hexane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:111-18-2
:
Formula:C₁₀H₂₄N₂
MDL Number:MFCD00008339
:172.31 g/mol
Boiling Pt:210°C
Melting Pt:–46 °C
Density:0,8064 g/cm³ (15 °C)
Flash Pt:75°C
EINECS:203-842-9
UN:2922
ADR:8,III
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together