

α,α,α,α',α',α'-Hexafluoro-p-xylene ≥99.0% (by GC)
: TCI
:433-19-2
:
:C₈H₄F₆
:MFCD00000402
:214.11 g/mol
:117°C
:3 °C
:1,3808 g/cm³ (25 °C)
:22°C
:207-086-0
:1993
:3,II
:GHS02
:GHS07
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together