Order Entry
Italy
ContactUsLinkComponent
2-(2,4-Dichlorophenyl)ethanol ≥97.0% (by GC)
2-(2,4-Dichlorophenyl)ethanol ≥97.0% (by GC)
Catalog # TCIAD2921-5G
Supplier:  TCI
CAS Number:  
undefined
2-(2,4-Dichlorophenyl)ethanol ≥97.0% (by GC)
Catalog # TCIAD2921-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :191.06 g/mol
CAS Number:81156-68-5
Formula:C₈H₈Cl₂O
MDL Number:MFCD00192010