
4,5-Dichloro-o-phenylenediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5348-42-5
:
Formula:C₆H₆Cl₂N₂
MDL Number:MFCD00007723
:177.03 g/mol
Boiling Pt:305 °C (1013 hPa)
Melting Pt:163 °C
EINECS:226-305-0
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together