Order Entry
Italy
ContactUsLinkComponent
 
 
 
3,5-Bis(trifluoromethyl)benzyl bromide ≥96.0% (by GC)
 :  TCI
 
undefined
 
 
3,5-Bis(trifluoromethyl)benzyl bromide ≥96.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorles to Yellow to Orange powder to lump to clear liquid
  • Purity (GC):
    min. 96.0 %

 

Frequently Bought Together