
3,4-Dimethoxybenzylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5763-61-1
:
Formula:C₉H₁₃NO₂
MDL Number:MFCD00008116
:167.21
Boiling Pt:156°C
Density:1,109 g/cm³ (20 °C)
EINECS:227-287-7
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White or Colorless to Yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together