


Benzyltrimethylammonium chloride ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56-93-9
:
Formula:C₁₀H₁₆ClN
MDL Number:MFCD00011782
:185.7 g/mol
Melting Pt:239°C
Density:1,07 g/cm³ (20 °C)
EINECS:200-300-3
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 % (calcd.on dried substance)
- Solubility in Water:almost transparency
- Drying loss:max. 1.0 %
Specification Test Results
Frequently Bought Together