
1,3-Diphenyl-2-thiourea ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:102-08-9
:
Formula:C₁₃H₁₂N₂S
MDL Number:MFCD00004921
:228.32 g/mol
Merck Index:14,03333
Boiling Pt:365 °C (1013 hPa)
Melting Pt:150…153 °C
Density:1,3205 g/cm³ (20 °C)
EINECS:203-004-2
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:150.0 to 153.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together