
1,1-Dimethylurea ≥98.0% (by total nitrogen basis)
Catalog # TCIAD0809-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point (Decomposition):178.0 to 184.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:88.11 g/mol
EINECS:209-957-0
:GHS07
CAS Number:598-94-7
Formula:C₃H₈N₂O
MDL Number:MFCD00007959