

2,2'-Phenyliminodiethanol ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:120-07-0
:
Formula:(C₆H₅)N(CH₂CH₂OH)₂
MDL Number:MFCD00002845
:181.23 g/mol
Boiling Pt:348 °C (1013 hPa)
Melting Pt:56…59 °C
Density:1,201 g/cm³ (60 °C)
Flash Pt:200 °C
Storage Temperature:Ambient
EINECS:204-368-5
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:55.0 to 59.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together