Order Entry
Italy
ContactUsLinkComponent
 
 
(4-Iodophenoxy)acetic acid ≥97.0% (by GC)
 :  TCI
 
undefined
 
(4-Iodophenoxy)acetic acid ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %

 

Frequently Bought Together