
1-Bromo-3,3-diphenylpropane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:20017-68-9
:
Formula:(C₆H₅)₂CHCH₂CH₂Br
MDL Number:MFCD00044829
:275.19 g/mol
Boiling Pt:145°C
Melting Pt:43 °C
Storage Temperature:Refrigerator
EINECS:243-467-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:39.0 to 43.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together