Order Entry
Italy
ContactUsLinkComponent
 
 
 
1-(1-Naphthyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
undefined
 
 
1-(1-Naphthyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    188.0 to 193.0 deg-C
  • Solubility in hot Acetone:
    almost transparency

 

Frequently Bought Together