
1,1,4,4-Tetraphenyl-1,3-butadiene ≥99.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1450-63-1
:
Formula:(C₆H₅)₂CCHCHC(C₆H₅)₂
MDL Number:MFCD00004766
:358.48 g/mol
Boiling Pt:556 °C (1013 hPa)
Melting Pt:207…209 °C
EINECS:215-914-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Melting point:195.0 to 205.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
Frequently Bought Together