Order Entry
Italy
ContactUsLinkComponent
 
L(-)-Proline ≥99.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
L(-)-Proline ≥99.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (HPLC) (CAD):
    min. 99.0 area%
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Specific rotation [a]20/D:
    -84.0 to -86.0 deg (C=4, H2O)
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together