Order Entry
Italy
ContactUsLinkComponent
(R,R)-(-)-3,6-Octanediol ≥96.0% (by GC)
(R,R)-(-)-3,6-Octanediol ≥96.0% (by GC)
Catalog # TCIAO0293-500MG
Supplier:  TCI
CAS Number:  
(R,R)-(-)-3,6-Octanediol ≥96.0% (by GC)
Catalog # TCIAO0293-500MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    49.0 to 53.0 deg-C
  • Specific rotation [a]20/D:
    -13.0 to -17.0 deg (C=1, Chloroform)

Specification Test Results

About this item

 :
 :146.23 g/mol
CAS Number:129619-37-0
Formula:C₈H₁₈O₂
MDL Number:MFCD01631087