

(±)-3,5-Dimethyl-1-hexyn-3-ol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:107-54-0
:
Formula:C₈H₁₄O
MDL Number:MFCD00008941
:126.2 g/mol
Boiling Pt:150°C
Density:0,859 g/cm³ (12 °C)
Flash Pt:57°C
EINECS:203-500-9
UN:1987
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together