
(±)-4-Bromomandelic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAB0609-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:116.0 to 119.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:231.05 g/mol
Merck Index:14,01424
Boiling Pt:344 °C (1013 hPa)
Melting Pt:115…118 °C
EINECS:230-085-1
:GHS07
CAS Number:6940-50-7
Formula:C₈H₇BrO₃
MDL Number:MFCD00004232