Order Entry
Italy
ContactUsLinkComponent
2-Acetyl-3-methylpyrazine ≥98.0% (by GC)
Irritant
2-Acetyl-3-methylpyrazine ≥98.0% (by GC)
Catalog # TCIAA1978-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-Acetyl-3-methylpyrazine ≥98.0% (by GC)
Catalog # TCIAA1978-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :136.15 g/mol
Boiling Pt:69°C
Density:1,11 g/cm³ (20 °C)
Flash Pt:80°C
EINECS:245-889-8
 :GHS07
CAS Number:23787-80-6
Formula:C₇H₈N₂O
MDL Number:MFCD00014612