
6-Hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16712-64-4
:
Formula:C₁₁H₈O₃
MDL Number:MFCD00060070
:188.18 g/mol
EINECS:240-759-7
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:246.0 to 250.0 deg-C
Specification Test Results
Frequently Bought Together