Order Entry
Italy
ContactUsLinkComponent
 
 
4-Chloro-1-naphthol ≥98.0% (by titrimetric analysis) for biochemical research
 :  TCI
 
undefined
 
4-Chloro-1-naphthol ≥98.0% (by titrimetric analysis) for biochemical research
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    118.0 to 122.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together