
4-Chloro-1-naphthol ≥98.0% (by GC, titration analysis)
Catalog # TCIAC0570-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:117.0 to 121.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:178.62 g/mol
Boiling Pt:307 °C (1013 hPa)
Melting Pt:123 °C
Density:1,2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-068-5
:GHS07
CAS Number:604-44-4
Formula:ClC₁₀H₆OH
MDL Number:MFCD00003974