Order Entry
Italy
ContactUsLinkComponent
Tetrapropylammonium chloride ≥97.0% (by titrimetric analysis)
Irritant
Tetrapropylammonium chloride ≥97.0% (by titrimetric analysis)
Catalog # TCIAT2106-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Tetrapropylammonium chloride ≥97.0% (by titrimetric analysis)
Catalog # TCIAT2106-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Drying loss:
    max. 3.0 %

Specification Test Results

About this item

 :
 :221.81 g/mol
Melting Pt:230°C
Density:1,0334 g/cm³ (3 °C)
EINECS:227-375-5
 :GHS07
CAS Number:5810-42-4
Formula:C₁₂H₂₈ClN
MDL Number:MFCD00038729