

Dodecyltrimethylammonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1119-94-4
:
Formula:C₁₅H₃₄BrN
MDL Number:MFCD00011767
:308.35 g/mol
Melting Pt:264…266 °C
EINECS:214-290-3
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together