1,2-Epoxycyclohexane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Epoxycyclohexane, Cyclohexene oxide
CAS Number:286-20-4
:
Formula:C₆H₁₀O
MDL Number:MFCD00005162
:98.14 g/mol
Boiling Pt:130°C
Melting Pt:–27 °C
Density:0,97 g/cm³ (20 °C)
Flash Pt:27°C
Storage Temperature:Ambient
EINECS:206-007-7
UN:1993
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together