

ω-Bromo-4-phenylacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:135-73-9
:
Formula:C₁₄H₁₁BrO
MDL Number:MFCD00000202
:275.14 g/mol
Boiling Pt:354 °C (1013 hPa)
Melting Pt:129…130 °C
Storage Temperature:Refrigerator
EINECS:205-217-6
UN:3261
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:125.0 to 129.0 deg-C
- Solubility in hot EtOH:almost transparency
Specification Test Results
Frequently Bought Together