
2'-Hydroxy-5'-methoxyacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:705-15-7
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00008731
:166.18 g/mol
Boiling Pt:281 °C (1013 hPa)
Melting Pt:113 °C
EINECS:211-882-3
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystaline
- Purity (GC):min. 98.0 %
- Melting point:47.0 to 50.0 deg-C
Specification Test Results
Frequently Bought Together