
Propyl-3,4,5-trihydroxybenzoate ≥98.0% (by titrimetric analysis)
: TCI
:121-79-9
:
:C₁₀H₁₂O₅
:MFCD00002196
:212.2 g/mol
:14,07859
:449 °C (1013 hPa)
:145…148 °C
:1,21 g/cm³ (20 °C)
:204-498-2
:GHS07
:Ambient
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:146.0 to 150.0 deg-C
- Drying loss:max. 1.0 %
Frequently Bought Together