Order Entry
Italy
ContactUsLinkComponent
 
 
 
4,4-Dimethyloctane ≥98.0% (by GC)
  TCIAD1216-1ML
 :  TCI
 :  
undefined
 
 
4,4-Dimethyloctane ≥98.0% (by GC)
  TCIAD1216-1ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :142.28 g/mol
 :161°C
 :–69 °C
 :0,733 g/cm³ (25 °C)
 :3295
 :3,III
 :GHS02
 :GHS08
 :Ambient
 :15869-95-1
 :C₁₀H₂₂
 :MFCD00048813