Order Entry
Italy
ContactUsLinkComponent
3,3-Dimethylheptane ≥98.0% (by GC)
Flammable
Health
3,3-Dimethylheptane ≥98.0% (by GC)
Catalog # TCIAD1206-1ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Health
3,3-Dimethylheptane ≥98.0% (by GC)
Catalog # TCIAD1206-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :128.26 g/mol
Boiling Pt:137°C
Density:0,7254 g/cm³ (20 °C)
Flash Pt:23°C
UN:1920
ADR:3,III
 :GHS02
 :GHS08
Storage Temperature:Ambient
CAS Number:4032-86-4
Formula:C₉H₂₀
MDL Number:MFCD00048811