Order Entry
Italy
ContactUsLinkComponent
 
 
 
2,6-Dimethyloctane ≥90.0% (by GC)
  TCIAD1212-1ML
 :  TCI
 :  
undefined
 
 
2,6-Dimethyloctane ≥90.0% (by GC)
  TCIAD1212-1ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 90.0 %

 

 

 :
 :142.28 g/mol
 :158°C
 :–101 °C
 :0,7313 g/cm³ (20 °C)
 :34°C
 :3295
 :3,III
 :GHS02
 :GHS08
 :Ambient
 :2051-30-1
 :C₁₀H₂₂
 :MFCD00042892