
2-Propanesulphonyl chloride ≥95.0% (by GC)
: TCI
:10147-37-2
:
:C₃H₇ClO₂S
:MFCD00007453
:142.61 g/mol
:83°C
:–47 °C
:1,273 g/cm³ (20 °C)
:86°C
:233-415-2
:3265
:8,III
:GHS05
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together