
Palmitoyl chloride ≥97.0% (by GC, titration analysis)
: TCI
:112-67-4
:
:C₁₆H₃₁ClO
:MFCD00000742
:274.87 g/mol
:200°C
:12 °C
:0,91 g/cm³ (20 °C)
:166°C
:Refrigerator
:203-996-7
:3265
:8,III
:GHS05
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Frequently Bought Together