Order Entry
Italy
ContactUsLinkComponent
(R)-(-)-3-Chloro-1,2-propanediol ≥96.0% (by GC)
Corrosive
Toxic
Health
(R)-(-)-3-Chloro-1,2-propanediol ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
Corrosive
Toxic
Health
(R)-(-)-3-Chloro-1,2-propanediol ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Brown clear liquid
  • Purity (GC):
    min. 96.0 %
  • Optical purity (GC):
    min. 98.0 ee%
  • Specific rotation [a]20/D:
    -7.0 to -10.0 deg (C=2, H2O)

Specification Test Results

Related Information

Frequently Bought Together