
2-Thiophenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16629-19-9
:
Formula:C₄H₃ClO₂S₂
MDL Number:MFCD00005426
Boiling Pt:93°C
Melting Pt:33°C
Storage Temperature:Refrigerator
EINECS:240-677-1
UN:3261
ADR:8,II
:182.65 g/mol
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together